Learn More
Selleck Chemical LLC 1-Hydroxy-2-naphthoic acid
SDP
Catalog No. 5000004643
Encompass_Preferred
1-Hydroxy-2-naphthoic acid is a xenobiotic metabolite produced by the biodegradation of phenanthrene by microorganisms
Catalog No. 50-000-04643 Supplier Selleck Chemical LLC Supplier No. S636425MG
1-Hydroxy-2-naphthoic acid is a xenobiotic metabolite produced by the biodegradation of phenanthrene by microorganisms