Learn More
Medchemexpress LLC 1-hydroxy-2-naphthoic acid | 86-48-6 | MFCD00003960 | >=97.0% | 188.18 | C11H8O3 | 25 G
SDP
Catalog No. 5000304156
Encompass_Preferred
1-Hydroxy-2-naphthoic acid (CAS 86-48-6) is an aromatic carboxylic acid used as a reagent and intermediate in organic synthesis and research. It is supplied as a solid (white to gray) and is also available in solution formats for convenient handling and reconstitution.
  • Used as a reagent and intermediate in small-molecule synthesis
  • Supplied as a solid and in DMSO solution formats for convenience
  • White to gray solid, suitable for handling and storage
  • High purity materials commonly available (supplier data often ≥97%)
  • Molecular formula C11H8O3 and molecular weight 188.18 aid formulation and calculations
Catalog No. 50-003-04156 Supplier Medchemexpress LLC Supplier No. 25GHYW016103
1-Hydroxy-2-naphthoic acid (CAS 86-48-6) is an aromatic carboxylic acid used as a reagent and intermediate in organic synthesis and research. It is supplied as a solid (white to gray) and is also available in solution formats for convenient handling and reconstitution.
  • Used as a reagent and intermediate in small-molecule synthesis
  • Supplied as a solid and in DMSO solution formats for convenience
  • White to gray solid, suitable for handling and storage
  • High purity materials commonly available (supplier data often ≥97%)
  • Molecular formula C11H8O3 and molecular weight 188.18 aid formulation and calculations