Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-hydroxy-2-naphthoic acid | 86-48-6 | MFCD00003960 | >=97.0% | 188.18 g/mol | C11H8O3 | 1 ML
SDP

Catalog No. 5000302132
Encompass_Preferred

1-Hydroxy-2-naphthoic acid is a naphthoic acid derivative used as a reference standard and research reagent in studies of polycyclic aromatic hydrocarbon (PAH) metabolism, analytical chemistry, and organic synthesis. It is supplied in laboratory-ready formats for analytical applications; consult the safety data sheet for handling, storage, and purity details.

  • Used as a research reagent and analytical standard.
  • Applicable for studies of polycyclic aromatic hydrocarbon (PAH) metabolism and organic synthesis.
  • Purity ≥97.0%.
  • Molecular formula C11H8O3; molecular weight 188.18 g/mol.
  • Available as a 10 mM solution for convenient analytical use.

Catalog No. 50-003-02132 Supplier Medchemexpress LLC Supplier No. HYW01610310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hydroxy-2-naphthoic acid is a naphthoic acid derivative used as a reference standard and research reagent in studies of polycyclic aromatic hydrocarbon (PAH) metabolism, analytical chemistry, and organic synthesis. It is supplied in laboratory-ready formats for analytical applications; consult the safety data sheet for handling, storage, and purity details.

  • Used as a research reagent and analytical standard.
  • Applicable for studies of polycyclic aromatic hydrocarbon (PAH) metabolism and organic synthesis.
  • Purity ≥97.0%.
  • Molecular formula C11H8O3; molecular weight 188.18 g/mol.
  • Available as a 10 mM solution for convenient analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.