Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-hydroxy-2-naphthoic acid | 86-48-6 | MFCD00003960 | >=97.0% | 188.18 g/mol | C11H8O3 | 50 G
SDP

Catalog No. 5000302142
Encompass_Preferred

1-Hydroxy-2-naphthoic acid (CAS 86-48-6) is an aromatic carboxylic acid used as a chemical standard and synthetic intermediate in research and analytical chemistry. It is encountered as a biodegradation metabolite of polycyclic aromatic hydrocarbons and is supplied for laboratory use.

  • High purity typically ≥97.0% suitable for analytical applications.
  • Molecular weight 188.18 g/mol and formula C11H8O3 for accurate calculations.
  • Soluble in alcohol, benzene, and diethyl ether; insoluble in cold water.
  • Commonly used as a reference standard and synthetic intermediate.
  • Stable solid with standard laboratory storage and handling requirements.

Catalog No. 50-003-02142 Supplier Medchemexpress LLC Supplier No. HYW01610350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hydroxy-2-naphthoic acid (CAS 86-48-6) is an aromatic carboxylic acid used as a chemical standard and synthetic intermediate in research and analytical chemistry. It is encountered as a biodegradation metabolite of polycyclic aromatic hydrocarbons and is supplied for laboratory use.

  • High purity typically ≥97.0% suitable for analytical applications.
  • Molecular weight 188.18 g/mol and formula C11H8O3 for accurate calculations.
  • Soluble in alcohol, benzene, and diethyl ether; insoluble in cold water.
  • Commonly used as a reference standard and synthetic intermediate.
  • Stable solid with standard laboratory storage and handling requirements.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.