Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1α-hydroxy-3-epi-vitamin D3 | 58028-00-5 | 99.3% | 400.64 | C27H44O2 | 1 MG
SDP

Catalog No. 5000215597
Encompass_Preferred

1α-Hydroxy-3-epi-vitamin D3 is a research-grade metabolite of 1α,25-dihydroxyvitamin D3 reported to potently suppress parathyroid hormone (PTH) secretion. It is supplied as a high-purity analytical standard for biological and analytical studies.

  • Natural metabolite of 1α,25-dihydroxyvitamin D3.
  • Reported potent suppressor of parathyroid hormone secretion.
  • Chemical formula C27H44O2 and molecular weight 400.64.
  • Purity 99.3% and typically supplied in milligram quantities.
  • Recommended storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-15597 Supplier Medchemexpress LLC Supplier No. HY10003A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1α-Hydroxy-3-epi-vitamin D3 is a research-grade metabolite of 1α,25-dihydroxyvitamin D3 reported to potently suppress parathyroid hormone (PTH) secretion. It is supplied as a high-purity analytical standard for biological and analytical studies.

  • Natural metabolite of 1α,25-dihydroxyvitamin D3.
  • Reported potent suppressor of parathyroid hormone secretion.
  • Chemical formula C27H44O2 and molecular weight 400.64.
  • Purity 99.3% and typically supplied in milligram quantities.
  • Recommended storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.