Learn More
Medchemexpress LLC 1-hydroxy-7-azabenzotriazole | 39968-33-7 | MFCD00210053 | 100g
SDP
Catalog No. 5000265978
Encompass_Preferred
HOAT is a commonly used peptide bond forming agent that may become an impurity in drug synthesis but is not mutagenic HOAT can be used for peptide fragment polycondensation and can also inhibit the racemization of peptide polycondensation[1]
Catalog No. 50-002-65978 Supplier Medchemexpress LLC Supplier No. HYZ0275100G
HOAT is a commonly used peptide bond forming agent that may become an impurity in drug synthesis but is not mutagenic HOAT can be used for peptide fragment polycondensation and can also inhibit the racemization of peptide polycondensation[1]