Learn More
Cayman Chemical 1-hydroxy VItamIn D2 1mg
SDP
Catalog No. 502955481
Encompass_Preferred
A synthetic prodrug of 1,25-dihydroxy vitamin D2; used to suppress the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; stimulates bone growth; inhibits the growth of certain blastomas and prostate cancer cells
Catalog No. 50-295-5481 Supplier Cayman Chemical Supplier No. 120771 MG
A synthetic prodrug of 1,25-dihydroxy vitamin D2; used to suppress the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; stimulates bone growth; inhibits the growth of certain blastomas and prostate cancer cells