Learn More
Cayman Chemical 1-hydroxy VItamIn D3 10mg
SDP
Catalog No. 502954927
Encompass_Preferred
A synthetic prodrug which is converted to the vitamin D receptor agonist 1,25-dihydroxy vitamin D3 in vivo; suppresses the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; supports bone growth; provides renal and cardiovascular protection during chronic kidney disease
Catalog No. 50-295-4927 Supplier Cayman Chemical Supplier No. 1208210 MG
A synthetic prodrug which is converted to the vitamin D receptor agonist 1,25-dihydroxy vitamin D3 in vivo; suppresses the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; supports bone growth; provides renal and cardiovascular protection during chronic kidney disease