Learn More
Cayman Chemical 1-hydroxy Vitamin D3, Purity: ≥95%, CAS: 41294-56-8, 1 mg
SDP
Catalog No. 502953018
Encompass_Preferred
A synthetic prodrug which is converted to the vitamin D receptor agonist 1,25-dihydroxy vitamin D3 in vivo; suppresses the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; supports bone growth; provides renal and cardiovascular protection during chronic kidney disease
Catalog No. 50-295-3018 Supplier Cayman Chemical Supplier No. 120821 MG
A synthetic prodrug which is converted to the vitamin D receptor agonist 1,25-dihydroxy vitamin D3 in vivo; suppresses the synthesis and secretion of parathyroid hormone in secondary hyperparathyroidism; supports bone growth; provides renal and cardiovascular protection during chronic kidney disease