Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxyanthraquinone 98.0+%, TCI America™
SDP

Catalog No. H03545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03545G 5 g
H035425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03545G Supplier TCI America Supplier No. H03545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129-43-1
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.215
MDL Number MFCD00058946
InChI Key BTLXPCBPYBNQNR-UHFFFAOYSA-N
PubChem CID 8512
ChEBI CHEBI:28877
IUPAC Name 1-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)O

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxyanthraquinone
EINECSNumber (4)-0703
RTECSNumber CB7178000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.