Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile 98.0+%, TCI America™
SDP

Catalog No. H13035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13035G 5 g
H13031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13035G Supplier TCI America Supplier No. H13035G

Chemical Identifiers

CAS 93413-76-4
Molecular Formula C15H19NO2
Molecular Weight (g/mol) 245.322
MDL Number MFCD06658142
InChI Key ASYJSBPNAIDUHX-UHFFFAOYSA-N
Synonym 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol
PubChem CID 9899621
IUPAC Name 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)C(C#N)C2(CCCCC2)O

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 245.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.