Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hydroxycyclohexyl phenyl ketone

Catalog No. 405612250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-561-2250G 250 g
40-561-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-561-2250G Supplier Sigma Aldrich Supplier No. 405612250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Hydroxycyclohexyl phenyl ketone

Specifications

CAS 947-19-3
Melting Point 47°C to 50°C
Boiling Point 175°C (15 mmHg)
Molecular Formula C13H16O2
Linear Formula HOC6H10COC6H5
MDL Number MFCD00059561
Quantity 250 g
Synonym (1-Hydroxycyclohexyl)phenylmethanone; 1-Benzoyl-1-hydroxycyclohexane; 1-Benzoylcyclohexanol
Molecular Weight (g/mol) 204.26
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.