Learn More
Strem, An Ascensus Company CAS# 2809-21-4. 25g. 1-Hydroxyethylidene-1,1-diphosphonic acid, min. 95% HEDP. MFCD00070585
SDP
Catalog No. 5090110243
Encompass
CAS# 2809-21-4. 25g. 1-Hydroxyethylidene-1,1-diphosphonic acid, min. 95% HEDP. MFCD00070585. Molecular Weight: 206.03. Molecular Formula: C(OH)(CH3)(PO3H2)2. Color/form: white pwdr. Strem# 15-2900. http://www.strem.com/catalog/v/15-2900/
Catalog No. 50-901-10243 Supplier Strem, An Ascensus Company Supplier No. 15290025G
CAS# 2809-21-4. 25g. 1-Hydroxyethylidene-1,1-diphosphonic acid, min. 95% HEDP. MFCD00070585. Molecular Weight: 206.03. Molecular Formula: C(OH)(CH3)(PO3H2)2. Color/form: white pwdr. Strem# 15-2900. http://www.strem.com/catalog/v/15-2900/