Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxyethylidenebis(phosphonic acid), 96%

Catalog No. AA4350209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4350209 10 g
AA4350218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4350209 Supplier Thermo Scientific Chemicals Supplier No. 04350209
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2809-21-4 | C2H8O7P2 | 206.027 g/mol

1-Hydroxyethylidenebis(phosphonic acid) acts as human protein tyrosine phosphatase inhibitor. It is also useful for photographic applications. It finds application to strengthen bone, osteoporosis treatment and to treat Paget's disease of bone. In addition, it is used in corrosion inhibition in circulating cool water systems, oil fields and low-pressure boilers with electric power, the chemical industry, metallurgy and fertilizers. It is also utilized for formulating reverse osmosis and thermal desalination anti-scaling agents. Furthermore, it is used in detergents, water treatment, cosmetics and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2809-21-4
Molecular Formula C2H8O7P2
Molecular Weight (g/mol) 206.027
MDL Number MFCD00070585
InChI Key DBVJJBKOTRCVKF-UHFFFAOYSA-N
Synonym etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl
PubChem CID 3305
ChEBI CHEBI:4907
IUPAC Name (1-hydroxy-1-phosphonoethyl)phosphonic acid
SMILES CC(O)(P(=O)(O)O)P(=O)(O)O

Specifications

Odor Odorless
Quantity 10 g
UN Number UN3261
Merck Index 14,3863
Solubility Information Soluble in water.
Formula Weight 206.03
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-552-8
RTECSNumber SZ8562100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.