Learn More
AdipoGen 1-Hydroxyibuprofen
SDP
Supplier:  AdipoGen CDXH0943M010
Encompass_Preferred
Chemical. CAS 53949-53-4. Formula C13H18O3. MW 222.28. The metabolite 1-Hydroxyibuprofen Ibuprofen Impurity L is a degradation product of Ibuprofen which is a non-steroidal anti-inflammatory drug NSAID used to relieve pain from various conditions such as headache, dental pain or menstrual cramps. Ibuprofen is an anti-inflammatory inhibitor targeting COX-1 and COX-2. Ibuprofen is metabolized in the human liver to a number of metabolites including 1-hydroxyibuprofen 1-OH-Ibu, 2-OH-Ibu and 3-OH-Ibu, respectively. 1-Hydroxyibuprofen was determined as the dominant compound in soils. This product can be used as a working standard or secondary analytical reference standard.
Catalog No. 50-256-0680