Learn More
Cayman Chemical 1-Hydroxyanthraquinone, Purity: ≥95%, CAS: 129-43-1, 25 g
SDP
Catalog No. 502949858
Encompass_Preferred
An anthraquinone; increases the percentage of isolated rat hepatocytes undergoing unscheduled DNA synthesis at 5 and 500 µM; dietary administration induces formation of cecum and colon adenocarcinomas, as well as hepatocellular carcinomas, in rats at 1%
Catalog No. 50-294-9858 Supplier Cayman Chemical Supplier No. 3290525 G
An anthraquinone; increases the percentage of isolated rat hepatocytes undergoing unscheduled DNA synthesis at 5 and 500 µM; dietary administration induces formation of cecum and colon adenocarcinomas, as well as hepatocellular carcinomas, in rats at 1%