Learn More
Medchemexpress LLC 1-hydroxyoctadecane (stearyl alcohol) | 112-92-5 | MFCD00002823 | 250g
SDP
Supplier:  Medchemexpress LLC HYY1809250G
Encompass_Preferred
1-Hydroxyoctadecane (Stearyl alcohol) is a long-chain fatty alcohol and excipient 1-Hydroxyoctadecane forms a closed oil film thereby reducing transepidermal water loss (TEWL) and promoting skin moisturization 1-Hydroxyoctadecane improves Imiquimod (HY-B0180)-induced psoriatic skin inflammation in mice[1]
Catalog No. 50-002-66190