Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxyphenazine 95.0+%, TCI America™
SDP

Catalog No. H02895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02895G 5 g
H0289100MG 100 mg
H02891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H02895G Supplier TCI America Supplier No. H02895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 528-71-2
Molecular Formula C12H8N2O
Molecular Weight (g/mol) 196.209
MDL Number MFCD00059692
InChI Key KLJWTLFECGZQCL-UHFFFAOYSA-N
PubChem CID 68249
IUPAC Name 5H-phenazin-1-one
SMILES C1=CC=C2C(=C1)NC3=CC=CC(=O)C3=N2

Specifications

Melting Point 158°C
Quantity 5 g
Formula Weight 196.21
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxyphenazine
RTECSNumber SG1647000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.