Learn More
Medchemexpress LLC 1-hydroxypyrene | 5315-79-7 | MFCD00044543 | 98.6% | 218.25 | C16H10O | 100 MG
SDP
Catalog No. 5000304050
Encompass_Preferred
1-Hydroxypyrene is an analytical standard and endogenous metabolite used to monitor exposure to polycyclic aromatic hydrocarbons (PAHs). It is applied in analytical and research workflows to quantify pyrene exposure in biological samples and to study aryl hydrocarbon receptor (AhR)-mediated effects.
  • Used as an analytical standard for detection and quantification of pyrene exposure.
  • Acts as an orally active aryl hydrocarbon receptor agonist relevant to biological studies.
  • Provided in lab-friendly solid and solution formats for flexibility in assays.
  • High purity suitable for analytical and reference applications.
  • Well-characterized molecular properties (C16H10O, MW 218.25) for reliable identification.
Catalog No. 50-003-04050 Supplier Medchemexpress LLC Supplier No. HYW014075100MG
1-Hydroxypyrene is an analytical standard and endogenous metabolite used to monitor exposure to polycyclic aromatic hydrocarbons (PAHs). It is applied in analytical and research workflows to quantify pyrene exposure in biological samples and to study aryl hydrocarbon receptor (AhR)-mediated effects.
  • Used as an analytical standard for detection and quantification of pyrene exposure.
  • Acts as an orally active aryl hydrocarbon receptor agonist relevant to biological studies.
  • Provided in lab-friendly solid and solution formats for flexibility in assays.
  • High purity suitable for analytical and reference applications.
  • Well-characterized molecular properties (C16H10O, MW 218.25) for reliable identification.