Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-hydroxypyrene | 5315-79-7 | MFCD00044543 | 98.6% | 218.26 g/mol | C16H10O | 1 G
SDP

Catalog No. 5000304046
Encompass_Preferred

Analytical standard of 1-hydroxypyrene (CAS 5315-79-7) intended as a reference material for qualitative and quantitative research methods. Common uses include method development, calibration, and environmental or biological analysis by chromatography and mass spectrometry.

  • Analytical reference for qualitative and quantitative methods.
  • Suitable for HPLC, GC, and MS applications.
  • Available as solid and solution formats, including 1 g solid.
  • Molecular formula C16H10O; molecular weight 218.26 g/mol.
  • Purity reported as 98.58% (rounded to 98.6%); consult certificate of analysis for lot-specific value.
  • For research use only; not for clinical or human use.

Catalog No. 50-003-04046 Supplier Medchemexpress LLC Supplier No. HYW0140751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard of 1-hydroxypyrene (CAS 5315-79-7) intended as a reference material for qualitative and quantitative research methods. Common uses include method development, calibration, and environmental or biological analysis by chromatography and mass spectrometry.

  • Analytical reference for qualitative and quantitative methods.
  • Suitable for HPLC, GC, and MS applications.
  • Available as solid and solution formats, including 1 g solid.
  • Molecular formula C16H10O; molecular weight 218.26 g/mol.
  • Purity reported as 98.58% (rounded to 98.6%); consult certificate of analysis for lot-specific value.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.