Learn More
Medchemexpress LLC 1-hydroxypyrene | 5315-79-7 | MFCD00044543 | 99.8% | 218.25 | C16H10O | 1 ML
SDP
Catalog No. 5000304045
Encompass_Preferred
1-Hydroxypyrene is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) intended for research and analytical applications. The compound (CAS 5315-79-7) has formula C16H10O and molecular weight 218.25, and is provided with high reported purity for use as an analytical standard.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity suitable for analytical applications.
  • Intended for research use only.
  • Storage recommendations provided for solvent and powder forms.
  • Suitable as a reference standard for pyrene exposure biomarker analysis.
Catalog No. 50-003-04045 Supplier Medchemexpress LLC Supplier No. HYW01407510MM/1ML
1-Hydroxypyrene is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) intended for research and analytical applications. The compound (CAS 5315-79-7) has formula C16H10O and molecular weight 218.25, and is provided with high reported purity for use as an analytical standard.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity suitable for analytical applications.
  • Intended for research use only.
  • Storage recommendations provided for solvent and powder forms.
  • Suitable as a reference standard for pyrene exposure biomarker analysis.