Learn More
Medchemexpress LLC 1-hydroxypyrene-d9 | 132603-37-3 | MFCD04972163 | 100.0% | 227.31 | C16HD9O | 1 MG
SDP
Catalog No. 5000302766
Encompass_Preferred
1-Hydroxypyrene-d9 is the deuterium-labeled analogue of 1-hydroxypyrene supplied as a reference standard for analytical and biomonitoring applications. It is used as an internal standard for LC-MS/MS quantitation to assess exposure to polycyclic aromatic hydrocarbons and to improve accuracy and recovery correction in environmental and toxicology analyses.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity for reliable quantitation.
  • Solid material, white to off-white, suitable for analytical workflows.
  • Intended for biomonitoring of PAH exposure in urine and environmental samples.
  • Available in small sizes optimized for analytical use.
Catalog No. 50-003-02766 Supplier Medchemexpress LLC Supplier No. HYW014075S1MG
1-Hydroxypyrene-d9 is the deuterium-labeled analogue of 1-hydroxypyrene supplied as a reference standard for analytical and biomonitoring applications. It is used as an internal standard for LC-MS/MS quantitation to assess exposure to polycyclic aromatic hydrocarbons and to improve accuracy and recovery correction in environmental and toxicology analyses.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity for reliable quantitation.
  • Solid material, white to off-white, suitable for analytical workflows.
  • Intended for biomonitoring of PAH exposure in urine and environmental samples.
  • Available in small sizes optimized for analytical use.