Learn More
Medchemexpress LLC 1-hydroxypyrene-d9 | 132603-37-3 | MFCD04972163 | 100.0% | 227.31 g·mol⁻¹ | C16HD9O | 10 MG
SDP
Catalog No. 5000304182
Encompass_Preferred
Deuterium-labeled 1-hydroxypyrene (d9) is an isotopically labeled internal standard used for quantitative analysis of 1-hydroxypyrene in biological and environmental samples by GC-MS or LC-MS. Supplied as a high-purity white to off-white powder, it provides consistent response and traceability for analytical and metabolic studies.
  • High isotopic and chemical purity for accurate quantification.
  • Suitable as an internal standard for GC-MS and LC-MS methods.
  • Solid powder form for convenient handling and spiking.
  • Storage at -20°C preserves long-term stability.
  • Available in small sizes tailored to analytical workflows.
Catalog No. 50-003-04182 Supplier Medchemexpress LLC Supplier No. HYW014075S10MG
Deuterium-labeled 1-hydroxypyrene (d9) is an isotopically labeled internal standard used for quantitative analysis of 1-hydroxypyrene in biological and environmental samples by GC-MS or LC-MS. Supplied as a high-purity white to off-white powder, it provides consistent response and traceability for analytical and metabolic studies.
  • High isotopic and chemical purity for accurate quantification.
  • Suitable as an internal standard for GC-MS and LC-MS methods.
  • Solid powder form for convenient handling and spiking.
  • Storage at -20°C preserves long-term stability.
  • Available in small sizes tailored to analytical workflows.