Learn More
Medchemexpress LLC 1-hydroxypyrene-d9 | 132603-37-3 | MFCD04972163 | 100.0% | 227.31 g/mol | C16HD9O | 5 MG
SDP
Catalog No. 5000304001
Encompass_Preferred
1-Hydroxypyrene-d9 is a deuterium-labeled analogue of 1-hydroxypyrene used as an internal standard and tracer in analytical assays for polycyclic aromatic hydrocarbons (PAHs). It is intended for research use and supplied as a white to off-white solid with high isotopic and chemical purity.
  • High isotopic enrichment and 99.95% purity.
  • Suitable as an internal standard for GC-MS, LC-MS, and NMR.
  • White to off-white solid, convenient for small-scale analytical use.
  • Stable when stored at -20°C as powder and under recommended solvent conditions.
  • Packaged in small quantities for trace-level quantitation.
Catalog No. 50-003-04001 Supplier Medchemexpress LLC Supplier No. HYW014075S5MG
1-Hydroxypyrene-d9 is a deuterium-labeled analogue of 1-hydroxypyrene used as an internal standard and tracer in analytical assays for polycyclic aromatic hydrocarbons (PAHs). It is intended for research use and supplied as a white to off-white solid with high isotopic and chemical purity.
  • High isotopic enrichment and 99.95% purity.
  • Suitable as an internal standard for GC-MS, LC-MS, and NMR.
  • White to off-white solid, convenient for small-scale analytical use.
  • Stable when stored at -20°C as powder and under recommended solvent conditions.
  • Packaged in small quantities for trace-level quantitation.