Learn More
Medchemexpress LLC HY-14791A 5mg Medchemexpress, (±)-Ibipinabant CAS:362519-49-1 Purity:>98%
SDP
Catalog No. 501969916
Encompass_Preferred
Medchemexpress, HY-14791A 5mg (±)-Ibipinabant CAS:362519-49-1 (±)-Ibipinabant ((±)-SLV319) is the racemate of SLV319. (±)-Ibipinabant ((±)-SLV319) is a potent and selective cannabinoid-1 (CB-1) receptor antagonist with an IC50 of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9916 Supplier Medchemexpress LLC Supplier No. HY14791A5MG
Medchemexpress, HY-14791A 5mg (±)-Ibipinabant CAS:362519-49-1 (±)-Ibipinabant ((±)-SLV319) is the racemate of SLV319. (±)-Ibipinabant ((±)-SLV319) is a potent and selective cannabinoid-1 (CB-1) receptor antagonist with an IC50 of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.