Learn More
Alkali Scientific 1" Cardboard Freezer Boxes, Fits 0.2mL PCR Tubes with 196-Place Divider
SDP
Catalog No. 502336085
Encompass_Preferred
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.
Catalog No. 50-233-6085 Supplier Alkali Scientific Supplier No. FS196
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.