Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Indanol, 98%

Catalog No. p-147933
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186110100 10 g Glass bottle
AC186110500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186110100 Supplier Thermo Scientific Chemicals Supplier No. 186110100
Only null left
Add to Cart

CAS: 6351-10-6 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-6-6351
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003797
InChI Key YIAPLDFPUUJILH-UHFFFAOYNA-N
Synonym 1-indanol, indanol, indan-1-ol, 1-hydroxyhydrindene, 1h-inden-1-ol, 2,3-dihydro, 1h-indenol, 2,3-dihydro, 1-hydroxyindan, 1-hydroxyindane, 1-indanole, --indanol
PubChem CID 22819
ChEBI CHEBI:16697
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES C1CC2=CC=CC=C2C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 50°C to 54°C
Color White to Yellow
Boiling Point 260°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 06, 574
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,benzene and chloroform,slightly soluble in petroleum ether
Formula Weight 134.18
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 1-Indanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-755-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.