Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Indanone

Catalog No. I230410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I230410G 10 g
I2304100G 100 g
I230425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I230410G Supplier Sigma Aldrich Supplier No. I230410G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Indanone

Specifications

CAS 83-33-0
Melting Point 38°C to 40°C (lit.)
Density 1.103 g/mL (at 25°C (literature))
Boiling Point 243°C to 245°C (lit.)
Molecular Formula C9H8O
Linear Formula C9H8O
MDL Number MFCD00003785
Quantity 10 g
Synonym 1-Oxoindane; alpha-Hydrindone
Molecular Weight (g/mol) 132.16
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.