Learn More
Selleck Chemical LLC 1-Indanone S9346-25MG
SDP
Catalog No. 5000025471
Encompass_Preferred
1-Indanone (Indan-1-one Alpha-Indanone) is an oxidation product of Indan a component of fuels solvents and varnishes It is part of the steroid hormone biosynthesis and arachidonic acid metabolism pathways
Catalog No. 50-000-25471 Supplier Selleck Chemical LLC Supplier No. S934625MG
1-Indanone (Indan-1-one Alpha-Indanone) is an oxidation product of Indan a component of fuels solvents and varnishes It is part of the steroid hormone biosynthesis and arachidonic acid metabolism pathways