Learn More
Medchemexpress LLC 1-Indanone | 83-33-0 | 132.16 | 10 G
SDP
Catalog No. 5000448909
Encompass_Preferred
1-Indanone is a biochemical reagent and organic compound utilized in life science research. This oxidation product is a component found in fuels, solvents, and varnishes. It is also involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Biochemical reagent for life science research
  • Organic compound
  • Oxidation product
  • Component of fuels, solvents, and varnishes
  • Participates in steroid hormone biosynthesis
  • Participates in arachidonic acid metabolism pathways
  • High purity (99.9%)
Catalog No. 50-004-48909 Supplier Medchemexpress LLC Supplier No. HYW00433310G
1-Indanone is a biochemical reagent and organic compound utilized in life science research. This oxidation product is a component found in fuels, solvents, and varnishes. It is also involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Biochemical reagent for life science research
  • Organic compound
  • Oxidation product
  • Component of fuels, solvents, and varnishes
  • Participates in steroid hormone biosynthesis
  • Participates in arachidonic acid metabolism pathways
  • High purity (99.9%)