Learn More
Medchemexpress LLC 1-Indanone | 83-33-0 | 132.16 | 100 G
SDP
Catalog No. 5000448792
Encompass_Preferred
1-Indanone is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is an oxidation product. This compound is also a component found in fuels, solvents, and varnishes. Additionally, it is involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used in life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • Functions as an oxidation product
  • Component of fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis pathways
  • Involved in arachidonic acid metabolism pathways
Catalog No. 50-004-48792 Supplier Medchemexpress LLC Supplier No. HYW004333100G
1-Indanone is a biochemical reagent suitable for life science research. It serves as a biological material or organic compound and is an oxidation product. This compound is also a component found in fuels, solvents, and varnishes. Additionally, it is involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used in life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • Functions as an oxidation product
  • Component of fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis pathways
  • Involved in arachidonic acid metabolism pathways