Learn More
Medchemexpress LLC 1-Indanone | 83-33-0 | 132.16 | 500 G
SDP
Catalog No. 5000448831
Encompass_Preferred
1-Indanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is an oxidation product and a component of fuels, solvents, and varnishes. It is also involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used as a biological material
  • Used as an organic compound for life science research
  • An oxidation product
  • Component of fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis pathways
  • Involved in arachidonic acid metabolism pathways
Catalog No. 50-004-48831 Supplier Medchemexpress LLC Supplier No. HYW004333500G
1-Indanone is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is an oxidation product and a component of fuels, solvents, and varnishes. It is also involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used as a biological material
  • Used as an organic compound for life science research
  • An oxidation product
  • Component of fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis pathways
  • Involved in arachidonic acid metabolism pathways