Learn More
Medchemexpress LLC 1-Indanone (α-Hydrindone) | 83-33-0 | 132.16 | 25 G
SDP
Catalog No. 5000449905
Encompass_Preferred
1-Indanone is a biochemical reagent that can be utilized as a biological material or organic compound for life science-related research. It is an oxidation product and a component found in fuels, solvents, and varnishes. Additionally, it plays a role in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used as a biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Acts as an oxidation product
  • A component in fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways
Catalog No. 50-004-49905 Supplier Medchemexpress LLC Supplier No. 25GHYW004333
1-Indanone is a biochemical reagent that can be utilized as a biological material or organic compound for life science-related research. It is an oxidation product and a component found in fuels, solvents, and varnishes. Additionally, it plays a role in steroid hormone biosynthesis and arachidonic acid metabolism pathways.
  • Used as a biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Acts as an oxidation product
  • A component in fuels, solvents, and varnishes
  • Involved in steroid hormone biosynthesis and arachidonic acid metabolism pathways