Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-2-methylbutane (stabilized with Copper chip) 95.0+%, TCI America™
SDP

Catalog No. I05305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05305G 5 g
I053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05305G Supplier TCI America Supplier No. I05305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 616-14-8
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.05
MDL Number MFCD01321177
InChI Key RHBHXHXNWHTGSO-UHFFFAOYNA-N
PubChem CID 141479
IUPAC Name 1-iodo-2-methylbutane
SMILES CCC(C)CI

Specifications

Boiling Point 148°C
Quantity 5 g
UN Number 1993
Formula Weight 198.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-methylbutane (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.