Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-3-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. I006325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I006325G Supplier TCI America Supplier No. I006325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 645-00-1
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007218
InChI Key CBYAZOKPJYBCHE-UHFFFAOYSA-N
Synonym 3-iodonitrobenzene, m-iodonitrobenzene, benzene, 1-iodo-3-nitro, 3-nitroiodobenzene, meta-iodonitrobenzene, 3-i-nitrobenzene, iodo-3-nitrobenzene, meta-nitroiodobenzene, 1-iodo-3-nitro-benzene, 3-nitro-iodobenzen
PubChem CID 12574
ChEBI CHEBI:67123
IUPAC Name 1-iodo-3-nitrobenzene
SMILES C1=CC(=CC(=C1)I)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 249.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Iodo-3-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.