Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iododecane, 98%, stabilized

Catalog No. AC330365000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330365000 500 g Glass Bottle
AC330360250 25 g Glass Bottle
AC330361000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC330365000 Supplier Thermo Scientific Chemicals Supplier No. 330365000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2050-77-3 | C10H21I | 268.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-77-3
Molecular Formula C10H21I
Molecular Weight (g/mol) 268.18
MDL Number MFCD00001085
InChI Key SKIDNYUZJPMKFC-UHFFFAOYSA-N
PubChem CID 16314
IUPAC Name 1-iododecane
SMILES CCCCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.2500g/mL
Boiling Point 124.0°C (11.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4845 to 1.4865
Linear Formula CH3(CH2)9I
Quantity 500 g
Beilstein 01,168
Specific Gravity 1.25
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
Stabilizer copper
Formula Weight 268.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iododecane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 218-104-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.