Learn More
Medchemexpress LLC 1-iodoethane-1,1,2,2,2-d5 | 6485-58-1 | MFCD00001092 | >98.0% | 161.00 g/mol | C2D5I | 500 MG
SDP
Catalog No. 5000283297
Encompass_Preferred
1-Iodoethane-1,1,2,2,2-d5 is the deuterium-labeled analogue of iodoethane supplied as a stable isotope reagent for research applications. It functions as an internal standard and tracer for analytical methods, and as a deuterated alkylation reagent in synthetic chemistry. For research use only.
Catalog No. 50-002-83297 Supplier Medchemexpress LLC Supplier No. HYW131797500MG
1-Iodoethane-1,1,2,2,2-d5 is the deuterium-labeled analogue of iodoethane supplied as a stable isotope reagent for research applications. It functions as an internal standard and tracer for analytical methods, and as a deuterated alkylation reagent in synthetic chemistry. For research use only.