Learn More
Medchemexpress LLC 1-iodoethane-1,1,2,2,2-d5 | 6485-58-1 | MFCD00001092 | 99.6% | 161.00 g/mol | C2D5I | 100 MG
SDP
Catalog No. 5000283296
Encompass_Preferred
1-Iodoethane-1,1,2,2,2-d5 is a deuterium-labeled ethyl iodide (C2D5I) used as an isotopically labeled tracer and internal standard for quantitative analysis in analytical chemistry. It is suitable for use in NMR, GC-MS, and LC-MS workflows and is supplied in small laboratory pack sizes for analytical applications.
  • Deuterium-labeled ethyl iodide (C2D5I) for isotope tracing.
  • 99.6% purity for high analytical confidence.
  • Molecular weight 161.00 g/mol; CAS 6485-58-1 for unambiguous identification.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-83296 Supplier Medchemexpress LLC Supplier No. HYW131797100MG
1-Iodoethane-1,1,2,2,2-d5 is a deuterium-labeled ethyl iodide (C2D5I) used as an isotopically labeled tracer and internal standard for quantitative analysis in analytical chemistry. It is suitable for use in NMR, GC-MS, and LC-MS workflows and is supplied in small laboratory pack sizes for analytical applications.
  • Deuterium-labeled ethyl iodide (C2D5I) for isotope tracing.
  • 99.6% purity for high analytical confidence.
  • Molecular weight 161.00 g/mol; CAS 6485-58-1 for unambiguous identification.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Available in small research pack sizes for laboratory use.