Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Iodohexane

Catalog No. 23828725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-828-725G 25 g
23-828-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-828-725G Supplier Sigma Aldrich Supplier No. 23828725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-iodohexane undergoes palladium-catalyzed cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.L]nonane to give tetradecane
  • Thermal reactions of 1-iodohexane on ni (100) single crystal surfaces has been studied using temperature-programmed desorption and x-ray photoelectron spectroscopy

Specifications

CAS 638-45-9
Density 1.437 g/mL (at 25°C (literature))
Boiling Point 179°C to 180°C (lit.)
Molecular Formula C6H13I
Refractive Index n20/D 1.492 (literature)
Linear Formula CH3(CH2)5 I
MDL Number MFCD00001102
Quantity 25 g
Synonym Hexyl iodide
Molecular Weight (g/mol) 212.07
Percent Purity ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.