Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodoisoquinoline 98.0+%, TCI America™
SDP

Catalog No. I07505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07505G 5 g
I075025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07505G Supplier TCI America Supplier No. I07505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19658-77-6
Molecular Formula C9H6IN
Molecular Weight (g/mol) 255.06
MDL Number MFCD00234494
InChI Key FDDBUIWRNBGXHB-UHFFFAOYSA-N
PubChem CID 640964
IUPAC Name 1-iodoisoquinoline
SMILES IC1=NC=CC2=CC=CC=C12

Specifications

Melting Point 75°C
Quantity 5 g
Formula Weight 255.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Iodoisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.