Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodopentane, 97%, stabilized

Catalog No. AC412475000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412475000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412475000 Supplier Thermo Scientific Chemicals Supplier No. 412475000

missing translation for 'validMainframeMsg'

CAS: 628-17-1 | C5H11I | 198.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-17-1
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.05
MDL Number MFCD00001100
InChI Key BLXSFCHWMBESKV-UHFFFAOYSA-N
Synonym n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
PubChem CID 12335
IUPAC Name 1-iodopentane
SMILES CCCCCI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 155.0°C to 157.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4940 to 1.4960
Linear Formula CH3(CH2)4I
Quantity 500 g
Beilstein 01, 133
Specific Gravity 1.51
Solubility Information Solubility in water: insoluble
Stabilizer copper
Formula Weight 198.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Iodopentane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fr
EINECSNumber 211-030-0
RTECSNumber SA2975600
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.