Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodopyrene 98.0+%, TCI America™
SDP

Catalog No. I09771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09771G 1 g
I0977200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09771G Supplier TCI America Supplier No. I09771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34244-15-0
Molecular Formula C16H9I
Molecular Weight (g/mol) 328.152
InChI Key CWNJSSNWLUIMDP-UHFFFAOYSA-N
PubChem CID 13555714
IUPAC Name 1-iodopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)I

Specifications

Melting Point 91°C
Color Yellow
Quantity 1 g
Formula Weight 328.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Iodopyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.