Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid

Catalog No. CDS013703 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013703 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013703 Supplier Sigma Aldrich Supplier No. CDS013703500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cyclohexanone dimethyl ketal reacts with trimethylsilane in the presence of trimethylsilyl triflate to form the corresponding ether
  • It can also undergo allylation and propargylation in the presence of indium to form the corresponding homoallylic or homopropargylic alcohol, respectively

Specifications

Molecular Formula C10H11N3O2
Linear Formula C10H11N3O2
Quantity 50 mL
MDL Number MFCD01570664
Molecular Weight (g/mol) 205.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.