Learn More
Medchemexpress LLC 1-isopropyl-4-nitrobenzene-d4 | 1219803-36-7 | 98.2% | 169.21 | C9H7D4NO2 | 10 MG
SDP
Catalog No. 5000283496
Encompass_Preferred
1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analog of 1-isopropyl-4-nitrobenzene supplied as an isotope-labeled standard for research and analytical applications. It is intended for use as a tracer or internal standard in quantitative studies, method development, and pharmacokinetic investigations. The compound has formula C9H7D4NO2, molecular weight 169.21, and is provided at high purity.
Catalog No. 50-002-83496 Supplier Medchemexpress LLC Supplier No. HYW142667S10MG
1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analog of 1-isopropyl-4-nitrobenzene supplied as an isotope-labeled standard for research and analytical applications. It is intended for use as a tracer or internal standard in quantitative studies, method development, and pharmacokinetic investigations. The compound has formula C9H7D4NO2, molecular weight 169.21, and is provided at high purity.