Learn More
Medchemexpress LLC 1-isopropyl-4-nitrobenzene-d4 | 1219803-36-7 | 98.2% | 169.21 g/mol | C9H7D4NO2 | 5 MG
SDP
Catalog No. 5000283084
Encompass_Preferred
1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analogue of 1-isopropyl-4-nitrobenzene supplied for research and analytical applications. It is formulated as a stable isotope reference material intended for use as an internal standard, tracer, or reference in mass spectrometry, NMR, and mechanistic studies, with high isotopic labeling and analytical purity.
Catalog No. 50-002-83084 Supplier Medchemexpress LLC Supplier No. HYW142667S5MG
1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analogue of 1-isopropyl-4-nitrobenzene supplied for research and analytical applications. It is formulated as a stable isotope reference material intended for use as an internal standard, tracer, or reference in mass spectrometry, NMR, and mechanistic studies, with high isotopic labeling and analytical purity.