Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-isopropyl-4-nitrobenzene-d4 | 1219803-36-7 | 98.2% | 169.21 g/mol | C9H7D4NO2 | 50 MG
SDP

Catalog No. 5000282917
Encompass_Preferred

1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analogue of 1-isopropyl-4-nitrobenzene supplied as a stable isotope reference standard for analytical and research applications. It is intended for use as a tracer or internal standard in mass spectrometry and related workflows; follow the SDS for handling and storage.

  • Deuterium-labeled compound for isotopic tracing and reference standard use.
  • Purity: 98.2% as reported by the supplier.
  • Molecular formula C9H7D4NO2 and molecular weight 169.21 g/mol.
  • Suitable for use as an internal standard in mass spectrometry applications.
  • Recommended storage: pure form at -20°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research quantities (for example, 50 mg).

Catalog No. 50-002-82917 Supplier Medchemexpress LLC Supplier No. HYW142667S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Isopropyl-4-nitrobenzene-d4 is the deuterium-labeled analogue of 1-isopropyl-4-nitrobenzene supplied as a stable isotope reference standard for analytical and research applications. It is intended for use as a tracer or internal standard in mass spectrometry and related workflows; follow the SDS for handling and storage.

  • Deuterium-labeled compound for isotopic tracing and reference standard use.
  • Purity: 98.2% as reported by the supplier.
  • Molecular formula C9H7D4NO2 and molecular weight 169.21 g/mol.
  • Suitable for use as an internal standard in mass spectrometry applications.
  • Recommended storage: pure form at -20°C; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small research quantities (for example, 50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.