Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylimidazole 97.0+%, TCI America™
SDP

Catalog No. I084525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I084525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. I084525G Supplier TCI America Supplier No. I084525G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 4532-96-1
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00234239
InChI Key IPIORGCOGQZEHO-UHFFFAOYSA-N
PubChem CID 10176113
IUPAC Name 1-propan-2-ylimidazole
SMILES CC(C)N1C=CN=C1

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 g
Formula Weight 110.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Isopropylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.