Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylpyrazole 98.0+%, TCI America™
SDP

Catalog No. I09991ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09991ML 1 mL
I09995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09991ML Supplier TCI America Supplier No. I09991ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18952-87-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD07468920
InChI Key ANMVTDVBEDVFRB-UHFFFAOYSA-N
PubChem CID 13469816
IUPAC Name 1-(propan-2-yl)-1H-pyrazole
SMILES CC(C)N1C=CC=N1

Specifications

Color Colorless
Boiling Point 142°C
Quantity 1 mL
UN Number 1993
Formula Weight 110.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Isopropylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.