Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Isothiocyanato-PEG4-alcohol | 1835759-69-7 | C9H17NO4S | 100 MG
SDP

Catalog No. 5000354276
Encompass_Preferred

1-Isothiocyanato-PEG4-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are innovative small molecules composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest. This unique design enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins, offering a promising approach for developing targeted therapy drugs.

  • Molecular weight: 235.30
  • Appearance: Colorless to light yellow liquid
  • Application: PROTAC linker for the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective degradation of target proteins

Catalog No. 50-003-54276 Supplier Medchemexpress LLC Supplier No. HY141246100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Isothiocyanato-PEG4-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are innovative small molecules composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest. This unique design enables PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins, offering a promising approach for developing targeted therapy drugs.

  • Molecular weight: 235.30
  • Appearance: Colorless to light yellow liquid
  • Application: PROTAC linker for the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective degradation of target proteins

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.