Learn More
Medchemexpress LLC 1-Isothiocyanato-PEG4-alcohol | 1835759-69-7 | C9H17NO4S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141246
Encompass_Preferred
1-Isothiocyanato-PEG4-alcohol is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs for research purposes. This compound helps to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52784